missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human AMPK A1/B1/G1, GST Tag, His Tag Recombinant Protein
Human AMPK A1/B1/G1, GST Tag, His Tag Recombinant Protein
1540.00 CHF - 4570.00 CHF
Spezifikation
Zugriffsnummer | NP_006242.4 I NP_006244.2 I NP_002724.1 |
---|---|
Zur Verwendung mit (Anwendung) | Kinase Assay |
Gen-Alias | AMPK A1/B1/G1 |
Kinase-Familie | CAMKL Kinase Family |
Spezies | Human |
Produktcode | Marke | Beschreibung | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Beschreibung | Preis | Menge & Verfügbarkeit | |||||
10393953
|
Invitrogen™
PV4672 |
10μg |
1540.00 CHF
1 Stück |
Verfügbar ab: 10-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15438673
|
Invitrogen™
PR8376C |
100μg |
4570.00 CHF
100 Mikrogramm |
Verfügbar ab: 10-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
AMP-activated protein kinase (AMPK) is a heterotrimeric complex that acts as sensor of cellular energy levels. The signaling cascades initiated by activating AMPK are critical to regulating metabolic events in the liver, skeletal muscle, heart, adipose tissue, and pancreas. An impairment in fuel metabolism that occurs in obesity is a factor leading to type 2 diabetes. The insulin resistance associated with type 2 diabetes is most profound at the level of skeletal muscle, the primary site of glucose and fatty acid utilization. Activation of AMPK is of interest for the treatment of type 2 diabetes.Spezifikation
NP_006242.4 I NP_006244.2 I NP_002724.1 | |
AMPK A1/B1/G1 | |
Human | |
GST Tag, His Tag | |
Ser⁄Thr Kinases: CAMK Group | |
Baculovirus | |
Recombinant, Full Length |
Kinase Assay | |
CAMKL Kinase Family | |
Recombinant | |
Storage Buffer: 50 mM Tris, pH 7.5, 150 mM NaCl, 0.02% Triton X-100, 0.5 mM EDTA, 2 mM DTT, 50% Glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Dry Ice | |
Kinases & Inhibitors | |
Liquid |