missing translation for 'onlineSavingsMsg'
Learn More

Ammoniumsulfat, 99 %, Thermo Scientific Chemicals

Catalog Number p-7077311
Click to view available options
Menge:
500 g
2.5 kg
7783-20-2
H8N2O4S
132.13
MFCD00003391
BFNBIHQBYMNNAN-UHFFFAOYSA-N
ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate
6097028
CHEBI:62946
Diazanium; Sulfat
N.N.OS(O)(=O)=O
7783-20-2
H8N2O4S
132.13
MFCD00003391
BFNBIHQBYMNNAN-UHFFFAOYSA-N
ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate
6097028
CHEBI:62946
Diazanium; Sulfat
N.N.OS(O)(=O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Ammonium sulfate
Schmelzpunkt 280.0°C
pH 4.5 to 5.5 (5% aq. soln.)
Menge 500 g
Strukturformel (NH4)2SO4
Merck Index 15,552
Löslichkeitsinformationen Solubility in water: 754g/L. Other solubilities: insoluble in alcohol and acetone
Formelmasse 132.14
Reinheit (%) 99%
Güte Biochemie

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'