Learn More
Ammoniumdiammintetrathiocyanatochromat(III)-Monohydrat, ACS,93.0 % min., Thermo Scientific Chemicals
Beschreibung
Ammonium diamminetetrathiocyanatochromate(III) monohydrate is used to precipitate primary and secondary amines as their ammonium salts. It is a color reagent and precipitant for mercury. It is also used as a precipitant for primary and secondary amines and certain amino acids. It is a reagent used for quantitative studies, including titrimetric, gravimetric, and spechtometric assays. Additionally, Reinecke salt has also been shown to determine the concentration of Dimenhydrinate using a spectrophotometric procedure. Other studies use Reinecke salt as a chemical actinometer to calculate the photodegradation of Manidipine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spezifikation
Chemischer Name oder Material | Ammonium diamminetetrathiocyanatochromate (III) monohydrate |
Farbe | Rot |
Physikalische Form | Pulver |
Menge | 25 g |
Strukturformel | NH4[Cr(NH3)2(SCN)4]·H2O |
UN-Nummer | UN3077 |
Merck Index | 14,8128 |
Löslichkeitsinformationen | Insoluble in dilute hydrochloric acid; Moderately soluble in water; Soluble in alcohol |
Formelmasse | 354.44 (336.42 Anhydrous) |
Reinheit (%) | ≥93.0% |
Mehr anzeigen |
RUO – Research Use Only