Learn More
Ammoniumchlorid kristallin MP Biomedicals
Artikelnummer. 11426502 Alle ansehen MP Biomedicals Produkte
Change view
Click to view available options
Menge:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge
11426502 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 11426502 Lieferant MP Biomedicals Lieferanten-Nr. 0215010980
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Spezifikation
Chemischer Name oder Material Ammonium Chloride
CAS 12125-02-9
Physikalische Form Crystalline
Menge 100 g
Summenformel ClH4N
MDL-Nummer MFCD00011420
Synonym ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid
InChI-Schlüssel NLXLAEXVIDQMFP-UHFFFAOYSA-N
SMILES N.Cl
IUPAC-Name Aminhydrochlorid
Molekulargewicht (g/mol) 53.49
PubChem CID 25517
ChEBI CHEBI:31206
Formelmasse 53.5