missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammoniumcarbonat, Puriss., Honeywell Fluka™
Click to view available options
Menge:
1 kg
2.5 kg
6 x 1 kg
6 x 2.5 kg
Beschreibung
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Spezifikation
Chemischer Name oder Material | Ammonium Carbonate |
Namenshinweis | Meets analytical specification of NF, Ph.Franc.,FCC |
Menge | 6 x 2.5 kg |
Strukturformel | (NH4)2CO3 |
Beilstein | 4253487 |
Formelmasse | 96.09g/mol |
Güte | puriss. |
Glührückstand | ≤0.05% (as SO4) |
Dichte | Vapor Density: 2.7 (vs air) |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'