Learn More
AMG 18 hydrochloride, Tocris Biosciences™
IRE1 Inhibitors
Marke:  Tocris Bioscience 6166/5
Encompass
Artikelnummer. 15997910
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Alternative Produkte anzeigen »
Beschreibung
Beschreibung
AMG 18 hydrochloride is a potent and selective IRE1α inhibitor (IC50 values are 13 and 99 nM for recombinant and cellular IRE1α, respectively). Exhibits selectivity for IRE1α in a panel of over 100 other kinases.
Spezifikation
Spezifikation
2250019-92-0
2-Chloro-N-[6-methyl-5-[[3-[2-[(3S)-3-piperidinylamino]-4-pyrimidinyl]-2-pyridinyl]oxy]-1-naphthalenyl]benzenesulfonamide hydrochloride