Learn More
Euro Protection™ Altaite High-Cut Safety Shoes
Euro Protection™ Altaite High-Cut Safety Shoes are comfortable, snug, and durable, and feature a composite toe cap for protection.
Marke:  Euro Protection™ 9ALTH42
Additional Details : Gewicht : 0.01000kg
 Lernen Sie weitere Varianten dieses
Artikelnummer. 12063666
  • 81.83 CHF / 1 Stück
Voraussichtliches Versanddatum: 12-08-2021
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
  • Padded collar and bellows tongue for snug fit.
  • Heel scuff cap
  • Shock-absorbing EVA midsole.
  • Full grain leather and bright mesh combination upper.
  • Breathable 3D mesh lining.
  • Removable, textile and foam insole.
Spezifikation
Spezifikation
Brown, Beige
Leather
Cemented, shock-absorbing EVA midsole, nitrile rubber outsole, non-metallic (composite fabric) puncture-resistant sole.
High
603 g per shoe in size 8
Breathable, 3D Mesh
42
Nitrile, EVA, Composite Fabric
Composite
EN ISO 20345 : 2011