Learn More
ANSELL™ AlphaTec™ Nitrile Gloves
Click to view available options
Größe:
7
8
9
10
11
Packungsgröße:
12 Paare
Beschreibung
- 8 mil or 0.2 mm thickness allows dexterity and tactility without a loss of durability
- NBR offering protection against light acids, greases, oils, and liquids
- Engineered fit to allow optimum balance of fit and flexibility both on its own or as an over glove
- The latex-free formulation of these gloves presents no risk of Type 1 allergies
- Long cuff extends to the forearm
Spezifikation
Spezifikation
| ANSI-Abriebleistung | ANSI 0 |
| Zertifizierungen/Konformität (Europa) | EN 421:2010|EN ISO 21420:2020|EN ISO 374-1:2016|EN ISO 374-5:2016|Food Contact|ISO 18889:2019 |
| Chlorinated | No |
| Anwendungen | Industriell |
| Farbe | Green |
| Finger Thickness (English) | 7,9 mil |
| Finger Thickness (Metric) | 0.2 mm |
| Latex Free | Ja |
| Material | Nitril |
| Palm Thickness (English) | 7,9 mil |
| Mehr anzeigen |