Learn More
Ansell Alphatec™ 4000 Avant2 Airline Ultrasonically Welded and Taped - Model 756-G02
Artikelnummer. 17120825
Click to view available options
Größe:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Packungsgröße:
3 Stück
Questo articolo non è restituibile. Consulta la politica di reso
Codice prodotto. 17120825
missing translation for 'mfr': Ansell GR40TA0075604G02
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Questo articolo non è restituibile. Consulta la politica di reso
Alphatec™ 4000 Avant2 Airline Ultrasonically Welded and Taped chemical protective suit's welded and taped seams offer the highest level of defense against liquids and particulates.
  • Advanced defenses: The AlphaTec™ 4000 AVANT2 AIRline Ultrasonically Welded and Taped - Model 756 protective suit was assessed against more than 200 chemicals, including chemical warfare agents
  • Enhanced protection: This chemical protective suit's welded and taped seams offer the highest level of defense against liquids and particulates
  • Added features: The AlphaTec™ 4000 AVANT2 AIRline Ultrasonically Welded and Taped - Model 756 also features a panoramic visor design, which ensures a good field of vision for the wearer
  • Reduced risk: In addition to a range of sleeve options, this protective body suit also comes with attached socks and an elasticated boot over flap to reduce chemical ingress risk
DESIGN FEATURES
  • Fully encapsulated air-fed suit that features a permanently built in AlphaTec AVANT STS V1 airline regulator.
  • Suit features include internal air-channel ventilation system double cuffs, finger and thumb loop.
  • Attached boots ends with anti-slip PVC sole version also available and emergency rip cord.
  • Attached glove options:. No glove with finger loops, G02(B) AlphaTec™ 02-100 or G05(D) AlphaTec™ 29-500.
  • Styles with codes ending in A756 include an optional CEJN airline connector, styles with codes ending in 0756 do not.
TRUSTED_SUSTAINABILITY
Specifica
Zertifizierung/Konformität CE CATEGORY III (Type 3, EN 1073-1, EN 1149-5)
Farbe Green
Zur Verwendung mit (Anwendung) Contaminated Environments, Chemical Handling, Filling, blending and charging of raw materials, Transferring liquids and solids, Opening and draining pumps valves or lines, Loading and unloading of proces equipment, Raw material sample collection, Concentration mixing and palletisation, Hazmat response, Assembly and disassembly of large pipes
Hood Type Fully Encapsulated
Material Multi-layer Barrier Laminate Fabric
Größe Large
Knöchelform Attached Socks, Elasticated Boot Over Flap
Verschlusstyp Zipper
Kragenform Hood with a panoramic visor
Einwegprodukt Reusable
Geschlecht Unisex
Flaschenhalstyp Hood with Panoramic Visor
Produkttyp Chemical Protective Suit
Schutzartklasse CE CATEGORY III (Type 3, EN 1073-1, EN 1149-5)
Saumtyp Ultrasonically Welded and Taped
Sterilität Non-sterile
Vedi altri risultati Mostra meno risultati
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato