Learn More
Ansell™ AlphaTec™ 3000 Ultrasonically Welded and Taped- Model 103
Artikelnummer. p-200137397 Alle ansehen Ansell Produkte
Click to view available options
Größe:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
Artikelnummer. 31114760
Marke: Ansell™ YE30T0010302
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Lightweight multi-layer fabric, an effective chemical barrier with dual-zip systems and double cuffs
  • The AlphaTec™ 3000 Ultrasonically Welded & Taped- Model 103 protective suit features multi-layered fabric that ensures an effective barrier against numerous inorganic chemicals and biohazards
  • This protective barrier is further reinforced by its double cuffs, dual-zip systems and double zip enclosure, which extends wear life
  • Highly visible bright yellow color, this protective outer garment remains particularly noticeable in low-lit environments, enhancing wearer and workplace safety
  • In addition, it complies with EN 1149-5 antistatic standards
TRUSTED_SUSTAINABILITY
Spezifikation
Produkttyp Chemical Protective Clothing
Größe Small
Geschlecht Unisex
Material Multi-layer Barrier Laminate Fabric
Farbe Yellow
Einwegprodukt Single-use
Taillenform Elastic
Knöchelform Elastic
Stulpenform Double, Elastic
Flaschenhalstyp Collared
Hood Type Standard Collar
Verschlusstyp Double Zipper
Saumtyp Welded, Taped
Sterilität Non-sterile
Widerstand Chemicals, Anti-static, Particle, Bloodborne Pathogens
Zertifizierungen/Konformität (Europa) CE CATEGORY III (Type 3-B, Type 4-B, Type 5-B, Type 6-B, EN 1073-2, EN 14126, EN 1149-5)
Latexfrei Yes
Anzahl Taschen None
Zur Verwendung mit (Anwendung) Transferring Liquids and Solids