Learn More
Ansell Alphatec™ 3000 Apron with Sleeves Ultrasonically Welded - Model 243-G01
Artikelnummer. p-200080418
Click to view available options
Größe:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Packungsgröße:
25 Stück
30 Stück
Artikelnummer. 16824940
Marke: Ansell YE30T0024302G01
Nur noch null auf Lager
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
This item is not returnable. View return policy
Alphatec™ 3000 Apron with Sleeves Ultrasonically Welded has rear velcro fastening at the neck.
DESIGN FEATURES:
  • Apron with sleeves with rear velcro fastening at the neck and attached AlphaTec™ 37-676
RECOMMENDED FOR:
  • Unexpected leakages spills or other releases
  • Loading and unloading trucks and vehicles
  • Loading trucks and vehicles
  • Product inspection
  • Cleaning of plant and machines
TRUSTED_SUSTAINABILITY
Specifications
Typ Chemical-resistant Apron
Größe Small
Material Multi-layer Barrier Laminate Fabric
Farbe Yellow
Einwegprodukt Reusable
Verschlusstyp Hook and Loop
Sterilität Non-sterile
Gewicht (metrisch) 80 gsm
Saumtyp Welded
Zertifizierungen/Konformität (Europa) CE CATEGORY III (Type PB [3]-B, Type PB [4]-B, EN 14126)