missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell™ AlphaTec™ 2000 Ts PLUS Stitched and Taped
Click to view available options
Größe:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Packungsgröße:
45 Stück
Beschreibung
Designed to allow perspiration to escape from the suit yet and withstand saturation of liquid chemicals.
- Filter 100% of particulates down to 0.01 microns in size
- Proven protective barrier to low concentration liquid chemicals, liquid and particulate biological hazards
- Comfortable to wear featuring moisture vapor permeable fabric to help reduce the risk of heat stress
- Optimized body fit, improving wearer comfort and safety
- Anti-static, tested according to EN 1149-5
- Ultra low linting, reduced risk of contamination in critical areas
- Silicone free, critical in spray painting applications
- Tunnelled elasticated hood, wrists and ankles to help minimize the risk of linting and cross contamination
Spezifikation
Spezifikation
| Produkttyp | Controlled Environments ESD Coveralls |
| Größe | Large |
| Geschlecht | Unisex |
| Material | Mikroporöses Polyethylen-Vlies-Laminat |
| Farbe | White |
| Einwegprodukt | Reusable |
| Taillenform | Elastic |
| Knöchelform | Elastisch |
| Stulpenform | Elastic |
| Flaschenhalstyp | Hooded |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction