Learn More
Ansell™ AlphaTec™ 2000 STANDARD-Gebunden
Artikelnummer. p-200004954
Click to view available options
Größe:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Packungsgröße:
30 Stück
Artikelnummer. 16884770
Marke: Ansell™ WH20B0015602
Nur noch null auf Lager
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
AlphaTec™ 2000 STANDARD Bound provides both protection and comfort with exceptional liquid and particulate protection. Ideal for a wide range of industrial applications with low hazard liquid chemical repellence, particle protection, protection from pesticides and barrier to biological agents.
Provides both protection and comfort with exceptional liquid and particulate protection.
  • Ideal for a wide range of industrial applications
  • Low hazard liquid chemical repellence, particle protection, protection from pesticides and barrier to biological agents.
  • Proven protective barrier to low concentration liquid chemicals, liquids, and particulate biological hazards
  • Comfortable to wear featuring moisture vapor permeable fabric to help reduce the risk of heat stress
  • Optimized body fit, improving wearer comfort and safety
  • Anti-static, tested according to EN 1149-5
  • Low linting to help reduce the risk of cross contamination in critical areas
  • Silicone free, critical in spray painting applications
TRUSTED_SUSTAINABILITY
Spezifikation
Produkttyp Controlled Environments ESD Coveralls
Größe Small
Geschlecht Unisex
Material Mikroporöses Polyethylen-Vlies-Laminat
Farbe White
Einwegprodukt Reusable
Taillenform Elastic
Knöchelform Elastisch
Stulpenform Elastic
Flaschenhalstyp Hooded
Hood Type 3-panel
Verschlusstyp Zipper
Saumtyp Bound
Sterilität Unsteril
Schutzartklasse CE-KATEGORIE III
Widerstand Chemikalien, Antistatisch, Partikel
Kragenform Haube
Zertifizierung/Konformität CE CATEGORY III (Type 5-B, Type 6-B, EN 1073-2, EN 14126, DIN 32781, EN 1149-5)
Anzahl Taschen Keine