Learn More
Ansell AlphaTec™ 1500 PLUS Stitched Coveralls, Model 111, White
Alle ansehen Ansell Produkte
Click to view available options
Größe:
4XL
5X-Large
6X-Large
Packungsgröße:
1 Stück
Beschreibung
- Specialized protection: The AlphaTec™ 1500 PLUS Stitched - Model 111 delivers light chemical resistance, guards against asbestos and filters 99.9% of particles over 3 microns in size, as well as brick and cement dust, light aerosols and mists
- Heightened breathability: This asbestos protection suit's air and water vapor permeability also reduces wearers' exposure to heat stress
- Enhanced fit and features: Wearer comfort is augmented by this protective body suit's optimal fit, while a three-piece hood offers additional protective coverage, finger loops offer a secure hold and red stitching ensures easy identification
- Certified antistatic capabilities: The AlphaTec™ 1500 PLUS Stitched - Model 111 is tested against EN 1149-5 antistatic standards
Spezifikation
Spezifikation
| Produkttyp | Schutzanzug mit Kapuze |
| Größe | 5X-Large |
| Geschlecht | Unisex |
| Material | Antistatisches SMS |
| Farbe | White |
| Einwegprodukt | Reusable |
| Taillenform | Elastisch |
| Knöchelform | Elastisch |
| Stulpenform | Fingerschlaufen |
| Flaschenhalstyp | Hooded |
| Mehr anzeigen |