Learn More
Ansell AlphaTec™ 1500 PLUS Stitched Coveralls, Model 103, White
Artikelnummer. 18070120 Alle ansehen Ansell Produkte
Click to view available options
Größe:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4XL
5X-Large
Packungsgröße:
1 Stück
Artikelnummer. 18070120
Marke: Ansell WH15S0010307
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Provides comfortable protection from hazardous particulates in critical environments.
    • Specialized protection: The AlphaTec 1500 PLUS Stitched - Model 103 protective suit is made from SMS material, protecting wearers against liquid spray and particulates, filtering 99.9% of particulates over 3 microns
    • Heightened breathability: Air and water vapor permeability help this protective outer garment minimize the risk of excessive heat stress and maximize wearer comfort
    • Assured antistatic capabilities: The AlphaTec 1500 PLUS Stitched - Model 103 is also tested against EN 1149-5 and AATC 76 antistatic standards
    • Enhanced fit and features: Its optimized fit ensures comfort, with additional features including a two-way zipper with resealable storm flap, finger loops and an elasticated waist, wrist and ankles
TRUSTED_SUSTAINABILITY
Spezifikation
Produkttyp Collared Coveralls
Größe 3X-Large
Geschlecht Unisex
Material Antistatisches SMS
Farbe White
Einwegprodukt Reusable
Taillenform Elastic at Back
Knöchelform Elastisch
Stulpenform Fingerschlaufen
Flaschenhalstyp Kragen
Hood Type Ohne Kapuze
Verschlusstyp 2-Weg-Reißverschluss mit Windschutzleiste
Saumtyp Genäht
Sterilität Unsteril
Schutzartklasse CE Kategorie III (Typ 5, Typ 6)
Widerstand Antistatisch, Chemikalien, Partikel
Kragenform Collar
Zertifizierungen/Konformität (Europa) EN ISO 13982-1:2004 + A1:2010∣EN 13034:2005 + A1:2009∣CE-Nummer∣EN 1073-2:2002∣EN 1149-5:2018∣EN ISO 13688:2013
Latexfrei Yes
Zur Verwendung mit (Anwendung) Body Surface checking, marking and touching up, painting or cleaning tools or robots, unpacking of bulk goods, loading and unloading of proces equipment, production line support and maintenance, assembly and inspection of components, asbestos related work, handing of powders, fiber glass/resin applications/ceramic fibres