missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALK [R1275Q] Recombinant Human Protein, Invitrogen™
Human ALK R1275Q, internal fragment, amino acids L1066-S1437 (as in NCBI/Protein entry NP_004295.2), R1275Q mutation, N-terminal GST-HIS6 fusion protein with a 3C cleavage site, expressed in Sf9 insect cells.
574.00 CHF - 8710.00 CHF
Spezifikation
Zur Verwendung mit (Anwendung) | Enzyme and Protein Activity Assay, Industrial and Applied Science, Kinase Assay, Kinase Biology, Kinase Proteins and Expression Reagents, Pharma and Biopharma, Protein Assays and Analysis, Protein Biology, Target and Lead Identification and Validation |
---|---|
Zusammensetzung | 50mM HEPES pH 7.5, 100mM NaCl, 5mM DTT, 15mM reduced glutathione, 20% glycerol |
Molekulargewicht | 70.4kDa |
Lagerungsbedingungen | Store at -68 to -85°C. Aliquot at first use to avoid multiple freeze-thaws. |
Proteinlänge | aa 1066-1437 |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15468833
|
Invitrogen
PV6169 |
10μg |
574.00 CHF
10 Mikrogramm |
Verfügbar ab: 03-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15478833
|
Invitrogen
PV6170 |
100μg |
1942.00 CHF
100 Mikrogramm |
Verfügbar ab: 03-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15488833
|
Invitrogen
PV6171 |
1mg |
8710.00 CHF
1 Milligramm |
Verfügbar ab: 03-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
- Validated in LanthaScreen™ Kinase Binding Assay
- Protocols available for simple use and optimized performance
- Multiple pack sizes available for convenient use
Spezifikation
Enzyme and Protein Activity Assay, Industrial and Applied Science, Kinase Assay, Kinase Biology, Kinase Proteins and Expression Reagents, Pharma and Biopharma, Protein Assays and Analysis, Protein Biology, Target and Lead Identification and Validation | |
70.4kDa | |
aa 1066-1437 | |
Recombinant |
50mM HEPES pH 7.5, 100mM NaCl, 5mM DTT, 15mM reduced glutathione, 20% glycerol | |
Store at -68 to -85°C. Aliquot at first use to avoid multiple freeze-thaws. | |
Human | |
GST Tag, His Tag |