missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALDH8A1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 21602-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
This protein belongs to the aldehyde dehydrogenases family of proteins. It plays a role in a pathway of 9-cis-retinoic acid biosynthesis in vivo. This enzyme converts 9-cis-retinal into the retinoid X receptor ligand 9-cis-retinoic acid, and has approximately 40-fold higher activity with 9-cis-retinal than with all-trans-retinal. Therefore, it is the first known aldehyde dehydrogenase to show a preference for 9-cis-retinal relative to all-trans-retinal. Two transcript variants encoding distinct isoforms have been identified for this gene.Spezifikation
ALDH8A1 | |
Polyclonal | |
Unconjugated | |
ALDH8A1 | |
Aldehyde dehydrogenase 12, ALDH12, ALDH8A1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
237320, 64577, 685750 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8BH00, Q9H2A2 | |
ALDH8A1 | |
ALDH8A1 Fusion Protein Ag16214 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |