missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALDH4A1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 11604-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
This protein belongs to the aldehyde dehydrogenase family of proteins. This enzyme is a mitochondrial matrix NAD-dependent dehydrogenase which catalyzes the second step of the proline degradation pathway, converting pyrroline-5-carboxylate to glutamate. Deficiency of this enzyme is associated with type II hyperprolinemia, an autosomal recessive disorder characterized by accumulation of delta-1-pyrroline-5-carboxylate (P5C) and proline. Two transcript variants encoding the same protein have been identified for this gene.Specifications
ALDH4A1 | |
Polyclonal | |
Unconjugated | |
ALDH4A1 | |
ALDH4, ALDH4A1, P5C dehydrogenase, P5CD, P5CDh, P5CDhL, P5CDhS | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
212647, 641316, 8659 | |
-20°C | |
Liquid |
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P0C2X9, P30038, Q8CHT0 | |
Aldh4a1 | |
ALDH4A1 Fusion Protein Ag2147 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |