Learn More
Abnova™ ALDH1B1 Recombinant Protein
Human ALDH1B1 full-length ORF recombinant protein with GST-tag at N-terminal
Marke: Abnova™ H00000219-P01.25ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
This protein belongs to the aldehyde dehydrogenases family of proteins. Aldehyde dehydrogenase is the second enzyme of the major oxidative pathway of alcohol metabolism. This gene does not contain introns in the coding sequence. The variation of this locus may affect the development of alcohol-related problems.
- Theoretical MW (kDa): 82.61
- Preparation method: In vitro wheat germ expression system
- Purification: Glutathione Sepharose 4 fast flow
- Storage buffer: 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer
Sequence: MLRFLAPRLLSLQGRTARYSSAAALPSPILNPDIPYNQLFINNEWQDAVSKKTFPTVNPTTGEVIGHVAEGDRADVDRAVKAAREAFRLGSPWRRMDASERGRLLNLLADLVERDRVYLASLETLDNGKPFQESYALDLDEVIKVYRYFAGWADKWHGKTIPMDGQHFCFTRHEPVGVCGQIIPWN
FPLVMQGWKLAPALATGNTVVMKVAEQTPLSALYLASLIKEAGFPPGVVNIITGYGPTAGAAIAQHMDVDKVAFTGSTEVGHLIQKAAGDSNLKRVTLELGGKSPSIVLADADMEHAVEQCHEALFFNMGQCCCAGSRTFVEESIYNEFLERTVEKAKQRKVGNPFELDTQQGPQVDKEQFERVLGYIQLGQKEGAKLLCGGERFGERGFFIKPTVFGGVQDDMRIAKEEIFGPVQPLFKFKKIEEVVERANNTRYGLAAAVFTRDLDKAMYFTQALQAGTVWVNTYNIVTCHTPFGGFKESGNGRELGEDGLKAYTEVKTVTIKVPQKNS
Best use within three months from the date of receipt of this protein.
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Spezifikation
AAH01619 | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
82.61 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ALDH1B1 | |
Human | |
Recombinant | |
Solution |
Antibody Production, Protein Array, ELISA, Western Blot | |
219 | |
ALDH1B1 (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Wheat Germ (in vitro) | |
ALDH5/ALDHX/MGC2230 | |
ALDH1B1 | |
Wheat Germ (in vitro) | |
GST |