missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human AKT3 (PKB gamma), His Tag Recombinant Protein
Human AKT3 (PKB gamma), His Tag Recombinant Protein
850.00 CHF
Spezifikation
Zugriffsnummer | NP_005456 |
---|---|
Zur Verwendung mit (Anwendung) | Kinase Assay |
Molekulargewicht | 60.4 kDa |
Gen-Alias | AKT3 (PKB gamma) |
Kinase-Familie | AKT Kinase Family |
Beschreibung
AKT3 (PKB gamma) has been shown to be stimulated by platelet-derived growth factor (PDGF), insulin, and insulin-like growth factor 1 (IGF1). It is involved in a wide variety of biological processes including cell proliferation, differentiation, apoptosis, tumorigenesis, glycogen synthesis and glucose uptake.Spezifikation
NP_005456 | |
60.4 kDa | |
AKT Kinase Family | |
Recombinant | |
Storage Buffer: 20mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.01% Triton X-100, 0.1mM Na3VO4, 2mM DTT, 20% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Dry Ice |
Kinase Assay | |
AKT3 (PKB gamma) | |
Human | |
His Tag | |
Ser⁄Thr Kinases: ACG Group | |
Baculovirus |