Learn More
Invitrogen™ AKT3 (PKB gamma) Recombinant Human Protein
AKT3 (PKB gamma) has been shown to be stimulated by platelet-derived growth factor (PDGF), insulin, and insulin-like growth factor 1 (IGF1).
Marke: Invitrogen™ PR4643B
Additional Details : Gewicht : 0.01000kg
Beschreibung
AKT3 (PKB gamma) is involved in a wide variety of biological processes including cell proliferation, differentiation, apoptosis, tumorigenesis, glycogen synthesis and glucose uptake.
Spezifikation
NP_005456 | |
20mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.01% Triton X-100, 0.1mM Na3VO4, 2mM DTT, 20% glycerol | |
100μg | |
RUO - research use only | |
AKT3 (PKB gamma) | |
Human | |
His Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
60.4kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
AKT Kinase Family | |
Recombinant | |
Baculovirus |