missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ AKR1B10 Recombinant Protein
Click to view available options
Menge:
10 μg
25 μg
Packungsgröße:
10 Mikrogramm
25 Mikrogramm
Beschreibung
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Spezifikation
Spezifikation
| Zugriffsnummer | AAH08837 |
| Zur Verwendung mit (Anwendung) | Antibody Production, Protein Array, ELISA, Western Blot |
| Zusammensetzung | 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer |
| Gen-ID (Entrez) | 57016 |
| Molekulargewicht | 60.5 |
| Name | AKR1B10 (Human) Recombinant Protein (P01) |
| pH-Bereich | 8 |
| Vorbereitungsmethode | In vitro wheat germ expression system |
| Reinigungsverfahren | Glutathione Sepharose 4 Fast Flow |
| Qualitätskontrollen | 12.5% SDS-PAGE Stained with Coomassie Blue. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction