Learn More
AIB-1 Mouse, Unlabeled, Clone: 34, BD
Mouse Monoclonal Antibody
370.00 CHF - 740.00 CHF
Spezifikation
Antigen | AIB-1 |
---|---|
Klon | 34 |
Konzentration | 250μg/mL |
Anwendungen | Western Blot |
Klassifikation | Monoclonal |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15835799
|
BD Biosciences
611105 |
150 μg |
740.00 CHF
1 Stück |
Verfügbar ab: 15-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15825799
|
BD Biosciences
611104 |
50 μg |
Dieser Artikel wurde eingestellt und ist leider nicht mehr verfügbar. Sehen Sie sich das Produkt an, um alternative Produktvorschläge zu sehen oder kontaktieren Sie unseren Technischen Support unter 056 618 41 11. |
||||||
Beschreibung
Signal transduction via nuclear hormone receptors is important for cell growth and differentiation, development, and homeostasis. Nuclear hormone receptors are ligand-activated transcription factors that modulate target gene expression. These ligand/receptor complexes also interact with transcriptional coactivators which enhance ligand-dependent transcription. Various classes of coactivators have been identified, including SRC-1 and its related proteins, such as TIF-2/GRIP-1, RIP140, AIB-1, and TIF-1α and -1β. AIB-1 (Amplified In Breast cancer-1) is also known as pCIP, RAC3, and TRAM-1. It interacts with estrogen receptor (ER) and is overexpressed in breast cancer biopsies and several breast and ovarian cancer cell lines. Similar to SRC-1 and TIF2, AIB-1 contains a basic helix-loop-helix (bHLH) domain followed by a PAS (Per/Arnt/Sim) region, serine and threonine rich regions, and a charged cluster. In addition, AIB-1 contains three copies of the conserved LXXLL motif which is critical to its interaction with the nuclear receptor. Thus, AIB-1 is a coactivator of nuclear receptors that may participate in the development of steroid-dependent cancers.
Spezifikation
AIB-1 | |
250μg/mL | |
Monoclonal | |
Mouse | |
Cell Biology | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
IgG1 | |
Affinity Purified | |
Amplified In Breast cancer-1; pCIP; RAC3; TRAM-1 |
34 | |
Western Blot | |
Unconjugated | |
RUO | |
Human, Mouse, Rat | |
Human AIB-1 aa. 376-389 | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.