missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AGFG2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 11919-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
This gene is a member of the HIV-1 Rev binding protein (HRB) family and encodes a protein with one Arf-GAP zinc finger domain, several phe-gly (FG) motifs, and four asn-pro-phe (NPF) motifs. This protein interacts with Eps15 homology (EH) domains and plays a role in the Rev export pathway, which mediates the nucleocytoplasmic transfer of proteins and RNAs. Alternatively spliced transcript variants have been described, but their biological validity has not been determined.Spezifikation
AGFG2 | |
Polyclonal | |
Unconjugated | |
AGFG2 | |
AGFG2, ArfGAP with FG repeats 2, HRBL, RAB R, RABR | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
231801, 304375, 3268 | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunofluorescence | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O95081, Q80WC7 | |
AGFG2 | |
AGFG2 Fusion Protein Ag2546 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Liquid |