Learn More
Aescin, 98%, Thermo Scientific™
Marke:  Thermo Scientific Alfa Aesar J66968.06
Artikelnummer. 15478549
  • ONLINE EXCLUSIVE PRICE
    313.00 CHF
    304.04 CHF / 5 Gramm
    Sparen 8.96 CHF
    2% Rabatt
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung
Beschreibung
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
It is used and studied as a vasoprotective anti-inflammatory, anti-edematous and anti-nociceptive agent.1 It may be used to study its pharmacokinetics, safety and efficacy as a chemosensitizer and modulator of NF-κ B cell signaling
Solubility
Soluble in methanol.
Notes
Desiccate at room temperature. Incompatible with oxidizing agents.
Videos
Sicherheitsdatenblatt (SDS) Dokumentation