Learn More
Sino Biological™ ADSL Recombinant Human Protein, N-His tag
Expressed from DNA sequence encoding human ADSL (Met 1-Leu 484) fused to polyhistidine tag at N-terminus
Marke: Sino Biological™ 11287H07E5
Additional Details : Gewicht : 0.10000kg
Beschreibung
Expressed from DNA sequence encoding human ADSL (Met 1-Leu 484) fused to polyhistidine tag at N-terminus
- Formulation: PBS, 10% Glycerol, pH 7.4
Spezifikation
Assay Standard | |
57kd | |
Human | |
<1 EU/μg | |
ADSL | |
His Tag | |
Other proteins | |
≥90% by SDS-PAGE |
158 | |
5μg | |
Recombinant | |
Adenylosuccinate Lyase | |
ADSL Recombinant Human Protein | |
E.coli | |
Lyophilized |
For Research Use Only. Not for use in diagnostic procedures.