missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADRBK2 (GRK3) Recombinant Human Protein
ADRBK2 (GRK3) Recombinant Human Protein
Marke: Invitrogen PV3827
Additional Details : Gewicht : 0.05250kg
Beschreibung
ADRBK2, or adrenergic beta receptor kinase 2 is a serine⁄threonine kinase that is thought to act as a regulator of receptor function.Spezifikation
NP_005151.2 | |
Pharma and Biopharma, Protein Assays and Analysis, Target and Lead Identification and Validation, Kinase Assay | |
107.2kDa | |
10μg | |
For research use only. Not for use in diagnostic procedures | |
ADRBK2 (GRK3) | |
ADRBK2 (GRK3) was pre-diluted in enzyme dilution buffer and assayed in 25mM Tris (pH 7.5), 10mM MgCl2, 0.5mM EGTA, 0.5mM Na3VO4, 5mM β-glycerophosphate, 2.5mM DTT, 0.01% Triton™ X-100, 10μM ATP, 100μg/mL GRKtide peptide substrate (RRREEEEESAAA) and trace [32P]-γ-ATP for 10 minutes at 30°C | |
ADRBK2 (GRK3) Recombinant Human Protein | |
Human | |
Baculovirus | |
Serine/threonine kinase |
0.38mg/mL | |
114294, 157 | |
ADRBK2 (GRK3) Recombinant Human Protein | |
–80°C | |
1 Each | |
ADRBK2 (GRK3) Recombinant Human Protein | |
GRK | |
1nmoles of phosphate transferred to GRKtide peptide substrate (RRREEEEESAAA) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 16.7μg/mL | |
GST Tag | |
Kinase and inhibitors | |
95% by SDS-PAGE |
For Research Use Only. Not for use in diagnostic procedures.