Learn More
Invitrogen™ Human ADRBK1 (GRK2), His Tag Recombinant Protein
Human ADRBK1 (GRK2), His Tag Recombinant Protein
Marke: Invitrogen™ PR4694C
Additional Details : Gewicht : 0.01000kg
Beschreibung
Many of the G protein-coupled receptors are known to be phosphorylated by G protein-dependent receptor kinases (GRKs)
Spezifikation
NP_001610.2 | |
82.3 kDa | |
ADRBK1 (GRK2) | |
Human | |
His Tag | |
Ser⁄Thr Kinases: ACG Group | |
Baculovirus | |
Recombinant, Full Length | |
Serine/Threonine Kinases |
Kinase Assay | |
100 μg | |
GRK Kinase Family | |
Recombinant | |
Storage Buffer: 50mM Tris-HCl, pH 7.5, 100mM NaCl, 0.5mM EDTA, 0.05% Triton X-100, 4mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Dry Ice | |
Kinases & Inhibitors | |
Liquid | |
Signal Transduction |
For Research Use Only. Not for use in diagnostic procedures.