missing translation for 'onlineSavingsMsg'
Learn More

Adenosin 5'-Monophosphatdinatriumsalz, TRC

Product Code. 17929104
Change view
Click to view available options
Menge:
10 g
Unit Size:
10 Gramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge Unit Size
17929104 10 g 10 Gramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17929104 Supplier Toronto Research Chemicals Supplier No. TRCA28179010G
 Eingeschränkter Artikel
Verfügbar ab: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Hochreine organische Moleküle und analytische Standards, strategisch weltweit bereitgestellt, um Innovation und kommerziellen Erfolg zu ermöglichen.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4578-31-8
Summenformel C10 H12 N5 O7 P . 2 Na
Molekulargewicht (g/mol) 391.18
Synonym 5'-Adenylic acid sodium salt (1:2), 5'-Adenylic acid, disodium salt (7CI,8CI,9CI), 5'-AMP disodium salt, AMP disodium salt, Adenosine 5'-monophosphate disodium, Adenosine 5'-monophosphate disodium salt, Adenosine 5'-phosphate disodium salt, Adenylic acid disodium salt, Disodium 5'-AMP, Disodium 9-(β-D-ribofuranosyl)adenine 5'-monophosphate, Disodium AMP, Disodium adenosine 5'-phosphate, Disodium adenylate
IUPAC-Name Dinatrium;[(2R,3S,4R,5R)-5-(6-Aminopurin-9-yl)-3,4-Dihydroxyoxolan-2-yl]methylphosphat
SMILES [Na+].[Na+].Nc1ncnc2c1ncn2[C@@H]3O[C@H](COP(=O)([O-])[O-])[C@@H](O)[C@H]3O

Specifications

Chemischer Name oder Material Adenosine 5'-Monophosphate Disodium Salt
Bezeichnung der Analyten oder Bestandteile Adenosine 5'-Monophosphate Disodium Salt
Versandbedingung Raumtemperatur
InChI-Formel InChI=1S/C10H14N5O7P.2Na/c11-8-5-9(13-2-12-8)15(3-14-5)10-7(17)6(16)4(22-10)1-21-23(18,19)20;;/h2-4,6-7,10,16-17H,1H2,(H2,11,12,13)(H2,18,19,20);;/q;2*+1/p-2/t4-,6-,7-,10-;;/m1../s1
Lösungstyp Unverdünnt
Industry Type Forensik
Menge 10 g
Formelmasse 391.027
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.