Learn More
Adeninhydrochlorid, 99,20 %, MP Biomedicals™
Artikelnummer. 11425032 Alle ansehen MP Biomedicals Produkte
Change view
Click to view available options
Menge:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge
11425032 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 11425032 Lieferant MP Biomedicals Lieferanten-Nr. 0219460880
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Spezifikation
Chemischer Name oder Material Adenine hydrochloride
CAS 6055-72-7
Farbe Weiß
Prozentgehaltsbereich 0.992
Summenformel C5H8ClN5O
MDL-Nummer MFCD11046306
Menge 100 g
Synonym 7h-purin-6-amine hydrochloride hydrate, adenine hydrochloride hydrate, adenine hydrochloride hemihydrate, 1h-purin-6-amine monohydrochloride hydrate 2:1, ade hydrate hydrochloride, purine-6-ylamine, chloride, hydrate, c5h5n5.hcl.h2o, adenine hydrochloride, adeninehydrochloridehemihydrate, vitamin b4 hydrate hydrochloride
InChI-Schlüssel MYRDTAUFFBYTHA-UHFFFAOYSA-N
SMILES O.Cl.NC1=C2NC=NC2=NC=N1
IUPAC-Name 7H-Purin-6-amin-Hydrathydrochlorid
Molekulargewicht (g/mol) 189.60
PubChem CID 24211900
Formelmasse 171.6
Physikalische Form Pulver