missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ACVR1 (ALK2), GST Tag Recombinant Protein

Artikelnummer. 10638703
Click to view available options
Menge:
10 μg
Packungsgröße:
1 Stück
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 10638703

Marke: Invitrogen™ PV4877

om dit product te kopen Registreer vandaag om een webaccount aan te maken

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Catalytic.

Activin A receptor, type I (ACVR1) belongs to the transforming growth factor-beta (TGF-beta) superfamily of structurally related signaling proteins. ACVR1 signals a specific transcriptional response in concert with activin type II receptors. The ACVR1 gene encodes activin A type I receptor, which signals a particular transcriptional response in concert with activin type II receptors. Mutations in ACVR1 have been linked to Fibrodysplasia Ossificans Progressiva (FOP), a rare genetic disorder characterized by the formation of bone in muscles, tendons, and other connective tissues, as well as Epicanthus.
TRUSTED_SUSTAINABILITY

Spezifikation

Zugriffsnummer Q04771
Konzentration See Label
Zur Verwendung mit (Anwendung) Kinase Assay
Zusammensetzung 50mM tris with 0.02% polysorbate 20, 50% glycerol, 150mM NaCl, 0.5mM EDTA, 2mM DTT and no preservative; pH 7.5
Gen-ID (Entrez) 90
Molekulargewicht 69.1 kDa
Name Human ACVR1 (ALK2), GST Tag
pH-Bereich 7.5
Reinigungsverfahren Purified
Menge 10 μg
Lagerungsbedingungen -80°C, Avoid Freeze/Thaw Cycles
Kennzeichnung RUO
Gen-Alias activin A receptor type 1; activin A receptor type I; activin A receptor, type 1; activin A receptor, type I; activin A receptor, type II-like kinase 2; activin receptor I precursor; activin receptor type I; Activin receptor type-1; activin receptor-like kinase 2; activin type I receptor; ACTRI; ActR-I; ActRIA; Acvr; ACVR1; ACVR1A; ACVRLK2; ALK2; Alk-2; ALK2-B; Alk8; ATVR1; D330013D15Rik; FOP; hydroxyalkyl-protein kinase; serine/threonine-protein kinase receptor; serine/threonine-protein kinase receptor R1; SKR1; TGF-B superfamily receptor type I; Tgfb1; Tsk7L; TSK-7L; TSRI; TSR-I; type I TGF B receptor
Gebräuchliche Bezeichnung ACVR1
Gensymbol ACVR1
Kinase-Familie STKR Kinase Family
Proteinlänge 145-509
Produkttyp Protein
Konjugat Unconjugated
Spezies Human
Rekombinant Recombinant
Proteinmarkierung GST-tag
Sequenz 145-509
Inhalt und Lagerung -80°C, Avoid Freeze/Thaw Cycles
Assay-Eingabe Biochemical binding/kinetic
Kinase-Gruppe Ser⁄Thr Kinases: TKL Group
Versandbedingung Dry Ice
Zieleintrag ACVR1
Zielgen activin A receptor, type I
Expressionssystem Insect cells
Proteinfamilie Kinases & Inhibitors
Proteinform Recombinant, Catalytic
Form Liquid
Protein-Subtyp Serine/Threonine Kinases
Forschungskategorie Signal Transduction
Mehr anzeigen Weniger anzeigen
Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt