missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACTN4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 19096-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
This antibody recognizes 100-105 kDa of endogenous ACTN4 protein in various lysates.
Alpha actinins belong to the spectrin gene superfamily which represents a diverse group of cytoskeletal proteins, including the alpha and beta spectrins and dystrophins. Alpha actinin is an actin-binding protein with multiple roles in different cell types. In nonmuscle cells, the cytoskeletal isoform is found along microfilament bundles and adherens-type junctions, where it is involved in binding actin to the membrane. In contrast, skeletal, cardiac, and smooth muscle isoforms are localized to the Z-disc and analogous dense bodies, where they help anchor the myofibrillar actin filaments. This gene encodes a nonmuscle, alpha actinin isoform which is concentrated in the cytoplasm, and thought to be involved in metastatic processes. Mutations in this gene have been associated with focal and segmental glomerulosclerosis.Spezifikation
ACTN4 | |
Polyclonal | |
Unconjugated | |
ACTN4 | |
ACTININ 4, actinin, alpha 4, ACTN4, Alpha actinin 4, F actin cross linking protein, FSGS, FSGS1, Non muscle alpha actinin 4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
60595, 63836, 81 | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
0.41 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O43707, P57780, Q9QXQ0 | |
Actn4 | |
ACTN4 Fusion Protein Ag13546 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Liquid |