Learn More
ACROBiosystems ActiveMax™ Human TNF-alpha Protein, Tag Free, low endotoxin (active trimer)
MALS verified
auf Anfrage - 1475.38 CHF
Spezifikation
Zugriffsnummer | NP_000585.2 |
---|---|
Molekulargewicht | 17.4 kDa |
Quelle | ActiveMax™ Human TNF-alpha, Tag Free, low endotoxin (active trimer) (MALS verified) (TNA-H4211) is expressed from human 293 cells (HEK293). It contains AA Val 77 - Leu 233 (Accession # P01375-1). |
Immunogen | Val 77 - Leu 233 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15897827
|
ACROBiosystems
TNA-H4211-100UG |
100 μg |
239.59 CHF
100 Mikrogramm |
Verfügbar ab: 16-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15807837
|
ACROBiosystems
TNA-H4211-1MG |
1 mg |
1475.38 CHF
1 Milligramm |
Verfügbar ab: 16-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15817837
|
ACROBiosystems
TNA-H4211-25UG |
25 μg |
Dieser Artikel wurde eingestellt und ist leider nicht mehr verfügbar. Sehen Sie sich das Produkt an, um alternative Produktvorschläge zu sehen oder kontaktieren Sie unseren Technischen Support unter 056 618 41 11. |
||||||
Beschreibung
Tumor necrosis factor alpha (TNFα) is a cytokine produced primarily by monocytes and macrophages. It is found in synovial cells and macrophages in the tissues.The primary role of TNFα is in the regulation of immune cells. TNFα is able to induce apoptotic cell death, to induce inflammation, and to inhibit tumorigenesis and viral replication. Dysregulation of TNFα production has been implicated in a variety of human diseases, including major depression, Alzheimer's disease and cancer. Recombinant TNFα is used as an immunostimulant under the INN tasonermin. TNFα can be produced ectopically in the setting of malignancy and parallels parathyroid hormone both in causing secondary hypercalcemia and in the cancers with which excessive production is associated.
This protein carries no "e;tag"e;. The protein has a calculated MW of 17.4 kDa. The protein migrates as 17 kDa under reducing (R) condition (SDS-PAGE).Spezifikation
NP_000585.2 | |
ActiveMax™ Human TNF-alpha, Tag Free, low endotoxin (active trimer) (MALS verified) (TNA-H4211) is expressed from human 293 cells (HEK293). It contains AA Val 77 - Leu 233 (Accession # P01375-1). | |
-20°C | |
TNF-alpha | |
Human | |
PBS, pH7.4 | |
HEK293 | |
ActiveMax™ Cytokines |
17.4 kDa | |
Val 77 - Leu 233 | |
DIF,TNF-alpha,TNFA,TNFSF2,cachexin,cachectin,TNFα | |
Unconjugated | |
Tag Free | |
Less than 0.1 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |