Learn More
ACROBiosystems ActiveMax™ Human Activin A / INHBA Protein, Tag Free, low endotoxin
low endotoxin
Marke: ACROBiosystems ACA-H421B-1MG
Additional Details : Gewicht : 0.00001kg
Beschreibung
Activin and inhibin are two closely related protein complexes that have almost directly opposite biological effects. Activin enhances FSH biosynthesis and secretion, and participates in the regulation of the menstrual cycle. Many other functions have been found to be exerted by activin, including roles in cell proliferation, differentiation, apoptosis, metabolism, homeostasis, immune response, wound repair, and endocrine function. Conversely inhibin down regulates FSH synthesis and inhibits FSH secretion.Activins are nonglycosylated homodimers or heterodimers of various β subunits (βA, βB, βC, and βE in mammals), while Inhibins are heterodimers of a unique α subunit and one of the β subunits. Activin A is a widely expressed homodimer of two βA chains. The βA subunit can also heterodimerize with a βB or βC subunit to form Activin AB and Activin AC, respectively. The 14 kDa mature human βA chain shares 100% amino acid sequence identity with bovine, feline, mouse, porcine, and rat βA.
This protein carries no "e;tag"e;. The protein has a calculated MW of 13.0 kDa. the protein migrates as 14 kDa under reducing (R) condition, and 27 kDa under non-reducing (NR) condition (SDS-PAGE).Spezifikation
AAH07858.1 | |
1 mg | |
Gly 311 - Ser 426 | |
Activin A,INHBA | |
Unconjugated | |
Tag Free | |
Less than 0.1 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
13.0 kDa | |
ActiveMax™ Human Activin A, Tag Free, low endotoxin (ACA-H421b) is expressed from human 293 cells (HEK293). It contains AA Gly 311 - Ser 426 (Accession # AAH07858.1). | |
-20°C | |
Activin A | |
Human | |
0.056% TFA in 60% ACN | |
HEK293 | |
ActiveMax™ Cytokines |