missing translation for 'onlineSavingsMsg'
Learn More

ACTH Mouse, Clone: SPM501, enQuireBio™

Mouse Monoclonal Antibody

Marke:  EnquireBio 5443-MSM7X-P1

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15910678

  • 505.00 CHF / 100 Mikrogramm


Verfügbar ab
25-03-2020
Zum Warenkorb hinzufügen

Spezifikation

Spezifikation

ACTH
Cytoplasmic
200μg/mL
10mM PBS with 0.05% BSA and 0.05% Azide
POMC
Synthetic peptide corresponding to aa25-39 of human ACTH (NGAEDESAEAFPLEF)
Normal pituitary gland or pituitary tumor
100 μg
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
Monoclonal
Immunohistochemistry
SPM501
Unconjugated
NP_001306133
Mouse
IgG1
Protein A/G Purified
RUO
Primary
5443
Videos
Dokumentation