missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ACSL4 Recombinant Rabbit Monoclonal Antibody (HL229)

Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA536264
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
ACSL4 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
The protein encoded by this gene is an isozyme of the long-chain fatty-acid-coenzyme A ligase family. Although differing in substrate specificity, subcellular localization, and tissue distribution, all isozymes of this family convert free long-chain fatty acids into fatty acyl-CoA esters, and thereby play a key role in lipid biosynthesis and fatty acid degradation. This isozyme preferentially utilizes arachidonate as substrate. The absence of this enzyme may contribute to the mental retardation or Alport syndrome. Alternative splicing of this gene generates 2 transcript variants.
Spezifikation
| ACSL4 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| O35547, O60488, Q9QUJ7 | |
| Acsl4 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human FACL4. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| HL229 | |
| Unconjugated | |
| Acsl4 | |
| 9430020A05Rik; Acs4; ACSL4; acyl-CoA synthetase 4; acyl-CoA synthetase long-chain family member 4; Arachidonate--CoA ligase; AU018108; Facl4; fatty acid Coenzyme A ligase, long chain 4; fatty acid-Coenzyme A ligase long chain 4; fatty acid-Coenzyme A ligase, long chain 4; fatty-acid-Coenzyme A ligase, long-chain 4; L MRX63; LACS 4; Lacs4; lignoceroyl-CoA synthase; long-chain acyl-CoA synthetase 4; long-chain fatty-acid-Coenzyme A ligase 4; long-chain-fatty-acid--CoA ligase 4; mACS4; MRX63; MRX68 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 113976, 2182, 50790 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur