Learn More
Sino Biological™ ACOX1 Recombinant Human Protein, N-His tag
Peroxisomal Acyl-Coenzyme A Oxidase 1 (ACOX1), recombinant human protein is supplied in frozen protein. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard,
Marke: Sino Biological™ 11266H07B25
Additional Details : Gewicht : 0.10000kg
Beschreibung
Peroxisomal Acyl-Coenzyme A Oxidase 1 (ACOX1), recombinant human protein is supplied in frozen protein. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.This recombinant protein was expressed from a DNA sequence encoding the human ACOX1 (AAH08767.1) (Met 1-Leu 660) fused to a polyhistidine tag at the N-terminus.
Activity: The activity of this protein has not been tested.
Reconstitution: No reconstitution is needed. Store aliquots under sterile conditions at -80°C. Thaw protein on ice before use and avoid repeated freeze-thaw cycles.
Formulation: 20 mM Tris, 500 mM NaCl, 20% Glycerol, 3mM DTT, pH 7.0.
Expiration Date: Expires one year from date of receipt when stored as instructed.
This protein is manufactured by Sino Biological Inc.
Spezifikation
Assay Standard | |
76.7 kDa | |
PAGE | |
Store at -80 °C | |
ACOX, MGC1198, PALMCOX, SCOX | |
ACOX1 | |
Recombinant | |
Insect | |
Frozen |
51 | |
acyl-CoA oxidase 1, palmitoyl | |
5 x 5μg | |
< 1 EU/μg | |
Includes 5 vials of protein in suspension buffer | |
Human | |
His Tag | |
Other Proteins | |
≥ 90% by SDS-PAGE |
For Research Use Only. Not for use in diagnostic procedures.