missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acinus Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marke: Novus Biologicals NBP1-76855
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Acinus Polyclonal specifically detects Acinus in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Spezifikation
| Acinus | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| Acinus, ACN, apoptotic chromatin condensation inducer 1, apoptotic chromatin condensation inducer in the nucleus, fSAP152, functional spliceosome-associated protein 152, KIAA0670DKFZp667N107 | |
| Rabbit | |
| 220 kDa | |
| 0.1 mg | |
| Apoptosis, Cancer, Chromatin Research, Tumor Suppressors | |
| 22985 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 1 mg/ml | |
| Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 5-20 μg/mL | |
| AAD56724 | |
| ACIN1 | |
| Antibody was raised against a 35 amino acid peptide near the center of human Acinus. The immunogen is located within amino acids 760 - 810 of Acinus. Amino Acid Squence: DTSENRPENDVPEPP | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur