missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACBD4 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 20941-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
ACBD4 is a member of the acyl-coenzyme A binding domain containing protein family. All family members contain the conserved acyl-Coenzyme A binding domain, which binds acyl-CoA thiol esters. They are thought to play roles in acyl-CoA dependent lipid metabolism.Spezifikation
ACBD4 | |
Polyclonal | |
Unconjugated | |
ACBD4 | |
ACBD4, HMFT0700 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
79777 | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.62 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8NC06 | |
ACBD4 | |
ACBD4 Fusion Protein Ag15045 | |
150 μL | |
Primary | |
Human | |
Liquid |