missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ABL2 (Arg) Recombinant Human Protein

ABL2 (Arg) is an Abl-related non-receptor protein tyrosine kinase.

Marke:  Invitrogen™ PR4795A

Additional Details : Gewicht : 0.01000kg

Artikelnummer. 15458423

  • 2605.00 CHF / 100 Mikrogramm
Verfügbar ab: 17-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

ABL2 (Arg) links cell surface receptors to cell proliferation and/or cytoskeletal reorganization.

Spezifikation

Spezifikation

NP_009298.1
50mM Tris, pH 7.5, 200mM NaCl, 2mM EDTA, 0.05% NP-40, 5mM DTT, 50% glycerol
100μg
RUO - research use only
ABL2 (Arg)
Human
His Tag
Frozen
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology
130.9kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
ABL Kinase Family
Recombinant
Baculovirus
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote