missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ABL1 [E255K] Recombinant Human Protein
The E255K mutant is a Gleevec resistant form of ABL1.
4170.00 CHF - 19680.00 CHF
Spezifikation
Zugriffsnummer | NP_005148.2, E255K |
---|---|
Zur Verwendung mit (Anwendung) | Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology |
Zusammensetzung | 50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% NP-40, 2mM DTT and 50% glycerol |
Molekulargewicht | 125.4kDa |
Lagerungsbedingungen | Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15458583
|
Invitrogen™
PR7420A |
100μg |
4170.00 CHF
100 Mikrogramm |
Verfügbar ab: 09-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
10251403
|
Invitrogen™
PV4182 |
1mg |
19680.00 CHF
1 Milligramm |
Verfügbar ab: 09-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
The ABL1 proto-oncogene encodes a cytoplasmic and nuclear protein tyrosine kinase that has been implicated in processes of cell differentiation, cell division, cell adhesion, and stress response.
Spezifikation
NP_005148.2, E255K | |
50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% NP-40, 2mM DTT and 50% glycerol | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
ABL Kinase Family | |
Recombinant | |
Baculovirus |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
125.4kDa | |
RUO - research use only | |
ABL1 E255K, ABL, c-ABL | |
Human | |
His Tag | |
Frozen |