missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AADAT Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 13031-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
This gene encodes a protein that is highly similar to mouse and rat kynurenine aminotransferase II. The rat protein is a homodimer with two transaminase activities. One activity is the transamination of alpha-aminoadipic acid, a final step in the saccaropine pathway which is the major pathway for L-lysine catabolism. The other activity involves the transamination of kynurenine to produce kynurenine acid, the precursor of kynurenic acid which has neuroprotective properties. Two alternative transcripts encoding the same isoform have been identified, however, additional alternative transcripts and isoforms may exist.Spezifikation
AADAT | |
Polyclonal | |
Unconjugated | |
AADAT | |
2 aminoadipate transaminase, AADAT, aminoadipate aminotransferase, KAT/AadAT, KAT2, KATII, Kynurenine aminotransferase II | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23923, 29416, 51166 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q64602, Q8N5Z0, Q9WVM8 | |
AADAT | |
AADAT Fusion Protein Ag3676 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |