Learn More
Thermo Scientific™ 96-Well Non-Skirted Plates, Low Profile Promotion
Low profile 96-well plates for use in PCR and qPCR applications
Marke: Thermo Scientific™ AB0700
Additional Details : Gewicht : 0.39700kg
Beschreibung
- Low profile to reduce dead space and to increase PCR efficiency
- Available with black alpha-numeric lettering
- Improved access for liquid handling
- Maximum fill volume of 0.2 mL
- Cut corner: H12
Spezifikation
Standard | |
Clear | |
96 |
8 x 12 | |
Virgin Polypropylene |