Learn More
Greiner Bio-One 96-Well Microplate CELLSTAR™, Cell Culture-Treated
Beschreibung
- For barcoded versions: Unique Code 128 barcode on all four sides of the microplate, consisting of a plate type-specific 3-letter prefix followed by a unique 7-digit serial number
- ANSI-compliant footprint for compatibility with automated systems
- Physical surface treatment for improved cell adhesion
- Enhanced cell attachment and higher proliferation rates to improve and accelerate cell expansion
- Free of detectable DNase, RNase, and human DNA
- Non-pyrogenic
- Non-cytotoxic
- High optical clarity for optimal microscopic examination
- Stackable design for efficient storage and handling
- Chimney well design allows each individual well to stand on its own, minimizing the risk of sample carryover and cross-contamination
- Manufactured under DIN ISO 9001 guidelines
- Supplied in a pack of 32
Spezifikation
Spezifikation
| Barcode | Includes Barcode |
| Format | Standard |
| Array | 8 x 12 |
| Farbe | Black, Clear Bottom |
| Produkttyp | Pre-barcoded Plate |
| Material | Polystyrene |
| Anzahl Wells | 96 |
| Sterilität | Sterile |
| Oberflächenbehandlung | Tissue Culture |
| Wellform | Flat Bottom (F-well) |