Learn More
9-Hydroxyellipticine Hydrochloride, TRC
Artikelnummer. 17973405
Change view
Click to view available options
Menge:
10 mg
Packungsgröße:
10 Milligramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
17973405 10 mg 10 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 17973405 Lieferant Toronto Research Chemicals Lieferanten-Nr. TRCH94968310MG
om dit product te kopen Registreer vandaag om een webaccount aan te maken
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemische Identifikatoren
CAS 52238-35-4
Summenformel C17 H14 N2 O . H Cl
Molekulargewicht (g/mol) 298.767
Synonym 5,11-Dimethyl-6H-pyrido[4,3-b]carbazol-9-ol Hydrochloride;
IUPAC-Name 5,11-dimethyl-6H-pyrido[4,3-b]carbazol-9-ol;hydrochloride
SMILES Cl.Cc1c2ccncc2c(C)c3c4cc(O)ccc4[nH]c13
Spezifikation
Chemischer Name oder Material 9-Hydroxyellipticine Hydrochloride
Menge 10 mg
InChI-Formel InChI=1S/C17H14N2O.ClH/c1-9-14-8-18-6-5-12(14)10(2)17-16(9)13-7-11(20)3-4-15(13)19-17;/h3-8,19-20H,1-2H3;1H
Formelmasse 298.087