missing translation for 'onlineSavingsMsg'
Learn More

9-Fluorenon-1-Carbonsäure, 98 %

Catalog Number p-7024997
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1573-92-8
C14H8O3
224.22
MFCD00011537
CBEFMGJHEKAMNI-UHFFFAOYSA-N
9-oxo-9h-fluorene-1-carboxylic acid, 9-fluorenone-1-carboxylic acid, 9-oxo-9h-fluorenecarboxylic acid, 9-oxofluorenecarboxylic acid, 9-fluorenone-1-carboxylicacid, 9h-fluorene-1-carboxylic acid, 9-oxo, maybridge1_002192, 9-oxo-1-fluorenecarboxylic acid, 9-oxo-9h-fluorene-1-carboxylicacid, 9h-fluorenecarboxylic acid, 9-oxo
74083
9-oxo-9H-fluorene-1-carboxylic acid
OC(=O)C1=CC=CC2=C1C(=O)C1=CC=CC=C21
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1573-92-8
C14H8O3
224.22
MFCD00011537
CBEFMGJHEKAMNI-UHFFFAOYSA-N
9-oxo-9h-fluorene-1-carboxylic acid, 9-fluorenone-1-carboxylic acid, 9-oxo-9h-fluorenecarboxylic acid, 9-oxofluorenecarboxylic acid, 9-fluorenone-1-carboxylicacid, 9h-fluorene-1-carboxylic acid, 9-oxo, maybridge1_002192, 9-oxo-1-fluorenecarboxylic acid, 9-oxo-9h-fluorene-1-carboxylicacid, 9h-fluorenecarboxylic acid, 9-oxo
74083
9-oxo-9H-fluorene-1-carboxylic acid
OC(=O)C1=CC=CC2=C1C(=O)C1=CC=CC=C21

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 9-Fluorenone-1-carboxylic acid
Schmelzpunkt 196°C to 198°C
Menge 1 g
Formelmasse 224.22
Reinheit (%) 98%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'