missing translation for 'onlineSavingsMsg'
Learn More

8-Bromo-7-(2-butyn-1-yl)-3,7-dihydro-3-methyl-1-[(4-methyl-2-quinazolinyl)methyl]-1H-purine-2,6-dione, TRC

Artikelnummer. 30004717
Change view
Click to view available options
Menge:
1 g
2.5 g
10 g
Unit Size:
1 Gramm
10 Gramm
2.50 Gramm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge unitSize
30004717 2.5 g 2.50 Gramm
30009194 10 g 10 Gramm
30009826 1 g 1 Gramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30004717 Supplier Toronto Research Chemicals Supplier No. TRCB6805102.5G

om dit product te kopen Registreer vandaag om een webaccount aan te maken

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 853029-57-9
Summenformel C20 H17 Br N6 O2
Molekulargewicht (g/mol) 453.29
Synonym 8-Bromo-7-(2-butyn-1-yl)-3,7-dihydro-3-methyl-1-[(4-methyl-2-quinazolinyl)methyl]-1H-purine-2,6-dione, 1H-Purine-2,6-dione, 8-bromo-7-(2-butynyl)-3,7-dihydro-3-methyl-1-[(4-methyl-2-quinazolinyl)methyl]- (9CI), 8-Bromo-7-(2-butyn-1-yl)-3,7-dihydro-3-methyl-1-[(4-methyl-2-quinazolinyl)methyl]-1H-purine-2,6-dione, 1-[(4-Methylquinazolin-2-yl)methyl]-3-methyl-7-(2-butyn-1-yl)-8-bromoxanthine
IUPAC-Name 8-bromo-7-but-2-ynyl-3-methyl-1-[(4-methylquinazolin-2-yl)methyl]purine-2,6-dione
SMILES CC#CCn1c(Br)nc2N(C)C(=O)N(Cc3nc(C)c4ccccc4n3)C(=O)c12

Specifications

Chemischer Name oder Material 8-Bromo-7-(2-butyn-1-yl)-3,7-dihydro-3-methyl-1-[(4-methyl-2-quinazolinyl)methyl]-1H-purine-2,6-dione
Hinweise zur Reinheitsqualität HPLC
Menge 2.5 g
InChI-Formel InChI=1S/C20H17BrN6O2/c1-4-5-10-26-16-17(24-19(26)21)25(3)20(29)27(18(16)28)11-15-22-12(2)13-8-6-7-9-14(13)23-15/h6-9H,10-11H2,1-3H3
Formelmasse 452.0596
Reinheit (%) >95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.