Learn More
6,8-Dibromo-3-cyclohexyl-3,4-dihydro-2(1H)-quinazolinone, Mikromol™
Product Code. 30049667
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30049667 100 mg 100mg
30052459 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30049667 Supplier Mikromol Supplier No. MM0065.05
 Restricted Item
Verfügbar ab: 26-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
6,8-Dibromo-3-cyclohexyl-3,4-dihydro-2(1H)-quinazolinone
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 6,8-Dibromo-3-cyclohexyl-3,4-dihydro-2(1H)-quinazolinone
CAS 21237-84-3
Analyte or Component Names 6,8-Dibromo-3-cyclohexyl-3,4-dihydro-2(1H)-quinazolinone
Shipping Condition Room Temperature
Molecular Formula C14 H16 Br2 N2 O
Certifications/Compliance ISO 9001
Synonym 6,8-Dibromo-3-cyclohexyl-3,4-dihydro-2(1H)-quinazolinone
InChI Formula InChI=1S/C14H16Br2N2O/c15-10-6-9-8-18(11-4-2-1-3-5-11)14(19)17-13(9)12(16)7-10/h6-7,11H,1-5,8H2,(H,17,19)
SMILES Brc1cc(Br)c2NC(=O)N(Cc2c1)C3CCCCC3
IUPAC Name 6,8-dibromo-3-cyclohexyl-1,4-dihydroquinazolin-2-one
Molecular Weight (g/mol) 388.1
Quantity 100 mg
Formula Weight 385.9629 g/mol