missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human IL-3 Protein
Click to view available options
Menge:
1 mg
10 μg
100 μg
250 μg
50 μg
500 μg
Packungsgröße:
1 Milligramm
10 Mikrogramm
100 Mikrogramm
250 Mikrogramm
50 Mikrogramm
500 Mikrogramm
Beschreibung
Measured in a cell proliferation assay using TF-1 human erythroleukemic cells. Kitamura, T. et al. (1989) J. Cell Physiol. 140:323. The ED50 for this effect is 0.02-0.1 ng/mL. The specific activity of Recombinant Human IL-3 is approximately 1.8 x 103 IU/μg, which is calibrated against recombinant human IL-3 WHO International Standard (NIBSC code: 91/510). Specific activity is for reference purposes only and is not routinely tested.
Spezifikation
Spezifikation
| Zugriffsnummer | AAC08706 |
| Zusammensetzung | Lyophilized from a 0.2um filtered solution in PBS. |
| Gen-ID (Entrez) | 3562 |
| Molekulargewicht | 15 kDa |
| Name | IL-3 |
| Menge | 500 μg |
| Quelle | E. coli-derived human IL-3 protein Ala20-Phe152,with and without an N-terminal Met |
| Lagerungsbedingungen | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Gen-Alias | Hematopoietic growth factor, IL3, IL-3MGC79398, interleukin 3 (colony-stimulating factor, multiple), interleukin-3, Mast cell growth factor, mast-cell growth factor, MCGF, MCGFMGC79399, MULTI-CSF, multilineage-colony-stimulating factor, Multipotential colony-stimulating factor, P-cell stimulating factor, P-cell-stimulating factor |
| Konjugat | Unconjugated |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur