Learn More
Thermo Scientific™ 5 L DynaDrive™ Bioreactor Probe Kit Puck Version
Artikelnummer. 31751204 Alle ansehen Thermo Scientific Produkte
Ansicht ändern
Click to view available options
Kapazität (metrisch):
5 L
Packungsgröße:
1 Stück
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Kapazität (metrisch) unitSize
31751204 5 L 1 Stück
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 31751204 Lieferant Thermo Scientific™ Lieferanten-Nr. F1102751001
 Eingeschränkter Artikel
Dieser Artikel erfordert eine Preisaktualisierung, bevor er bestellt werden kann.
Fordern Sie hier einen Preis für diesen Artikel an.
DynaDrive bioreactor puck sensor probe kit. Compatible with HyPerforma G3Lab controllers and 5 L DynaDrive bioreactor stands and containers using sensor puck technology. Enables pH and DO monitoring for bioprocess applications.
The Thermo Scientific™ DynaDrive™ 5 L BPC Sensor Puck Kit, F110-2751-001, enables rapid pH and dissolved oxygen (DO) measurement when using sensor puck–configured DynaDrive™ 5 L BioProcess Container (BPC). Designed for compatibility with Thermo Scientific™ HyPerforma™ G3Lab™ Bioprocess Controllers, this kit supports rapid setup, reduced handling, and reliable process monitoring during upstream process development.
The sensor puck kit integrates with the DynaDrive™ 5 L Single-Use Bioreactor ecosystem.
Key features and benefits
  • pH and DO measurement
  • Rapid setup
  • Designed specifically for use with sensor puck–enabled DynaDrive™ 5 L BPC
  • Compatible for use with HyPerforma™ G3Lab™ Bioprocess Controllers to support bioprocess control strategies
Kit includes
  • Puck Sensor Reader (Electro-Optical Module)
  • Cables for Puck Sensor Reader to connect to G3Lab controller
Typical applications
  • Upstream process development and optimization
  • Media and feed screening
  • Scale-down modeling for fed-batch or perfusion processes
Designed for the DynaDrive™ ecosystem
The F110-2751-001 sensor puck kit is part of the Thermo Scientific™ DynaDrive™ Single-Use Bioreactor platform, supporting efficient, scalable, and standardized upstream workflows from early process development through technology transfer.
TRUSTED_SUSTAINABILITY
Spezifikation
Kapazität (metrisch) 5 L
Material Stainless Steel (304)
Typ Probe Kit